missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FPRP/PTGFRN Antibody (1F11) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-21497JF635
This item is not returnable.
View return policy
Description
FPRP/PTGFRN Monoclonal antibody specifically detects FPRP/PTGFRN in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Frozen)
Specifications
| FPRP/PTGFRN | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CD315, CD9P-1, FPRP, PTGFRN | |
| HeLa cell line | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Frozen) | |
| 1F11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 5738 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction