missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRA-1/FOSL1 Antibody (OTI12F9), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71351G
This item is not returnable.
View return policy
Description
FRA-1/FOSL1 Monoclonal specifically detects FRA-1/FOSL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| FRA-1/FOSL1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FOSL1 | |
| Full-length protein expressed in 293T cell transfected with human FRA-1/FOSL1 expression vector. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI12F9 | |
| Western Blot, Immunohistochemistry | |
| FOS-like antigen 1, FOS-like antigen-1, fos-related antigen 1, FRA, fra-1, FRA1 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Wnt Signaling Pathway | |
| 8061.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction