missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRA-1/FOSL1 Antibody (OTI12F9), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71351PE
This item is not returnable.
View return policy
Description
FRA-1/FOSL1 Monoclonal antibody specifically detects FRA-1/FOSL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| FRA-1/FOSL1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI12F9 | |
| Western Blot, Immunohistochemistry | |
| FOS-like antigen 1, FOS-like antigen-1, fos-related antigen 1, FRA, FRA1, FRA-1 | |
| Full-length protein expressed in 293T cell transfected with human FOSL1 expression vector | |
| 0.1 mL | |
| Cell Cycle and Replication, Wnt Signaling Pathway | |
| 8061 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction