Learn More
Novus Biologicals™ FRA10AC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30678
Product Code. 30045074
  • € 571.00 / 100µg
Description
FRA10AC1 Polyclonal antibody specifically detects FRA10AC1 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FRA10AC1
Polyclonal
Tris-citrate/phosphate pH 7 -8.
Rabbit
Affinity purified
RUO
118924
Store at 4°C. Do not freeze.
IgG
Western Blot
Unconjugated
C10orf4fragile site 10q23.3, chromosome 10 open reading frame 4, F26C11.1-like, FRA10A, fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate1, protein FRA10AC1, rare folic acid-type fragile site, FRA(10)(q23.3), candidate gene 1
Between 265 and 315
100 μg
Primary
Human, Mouse