missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRA2 (FOSL2) Monoclonal Antibody (OTI1G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809658
This item is not returnable.
View return policy
Description
FRA2 (FOSL2) Monoclonal antibody specifically detects FRA2 (FOSL2) in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| FRA2 (FOSL2) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q5PQL4, Q92989, Q99LI9 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10978, 311166, 98985 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1G1 | |
| Unconjugated | |
| CLP1 | |
| FOS like 2, AP-1 trancription factor subunit; FOS like 2, AP-1 transcription factor subunit; FOS like antigen 2; FOSL2; FOS-like antigen 2; fos-related antigen 2; Fra2; FRA-2 | |
| Human recombinant protein fragment corresponding to amino acids 1-226 of human CLP1 (NP_006822) produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction