missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Francisella tularensis ssp. tularensis antigen FTT0571 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-62179
This item is not returnable.
View return policy
Description
Francisella tularensis ssp. tularensis antigen FTT0571 Polyclonal antibody specifically detects Francisella tularensis ssp. tularensis antigen FTT0571 in Bacteria, Virus samples. It is validated for Western Blot, ELISA
Specifications
| Francisella tularensis ssp. tularensis antigen FTT0571 | |
| Polyclonal | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Uncharacterized protein | |
| Recombinant FTT0571 protein produced in E. coli | |
| 0.1 mg | |
| Primary | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| PBS (pH 7.4) | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Bacteria, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction