missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRAS1 (Fraser syndrome 1) Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18434V
This item is not returnable.
View return policy
Description
FRAS1 (Fraser syndrome 1) Polyclonal antibody specifically detects FRAS1 (Fraser syndrome 1) in Human samples. It is validated for Western Blot
Specifications
| FRAS1 (Fraser syndrome 1) | |
| Polyclonal | |
| Western Blot | |
| DKFZp686I05113, extracellular matrix protein FRAS1, FLJ14927, FLJ22031, Fraser syndrome 1, KIAA1500DKFZp686P08111 | |
| Recognizes region between residues Between Between 3025 and 3075.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 80144 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction