missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frataxin Antibody (OTI1C12), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70766CL1
This item is not returnable.
View return policy
Description
Frataxin Monoclonal antibody specifically detects Frataxin in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| Frataxin | |
| Monoclonal | |
| CoraFluor 1 | |
| CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199 | |
| Full length human recombinant protein of human FXN(NP_000135) produced in HEK293T cell. | |
| 0.1 mL | |
| Diabetes Research | |
| 2395 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI1C12 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction