Learn More
Euro Protection™ Freedite Low-Cut Safety Shoes
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Description
- Footbridge for torsional rigidity.
- Padding for snug fit.
- Also available with an electrical hazards resistant outsole on request (Reference: 9FEHH).
- Full grain leather upper, reflective trim at the back.
- Breathable 3D mesh lining.
- Removable, textile and foam insole.
- Composite toe cap.
- Non-metallic (composite fabric) puncture resistant sole.
- Injected polyurethane midsole.
- Polyurethane outsole.
Specifications
Specifications
| Color | Black |
| Liner Material | Breathable 3D mesh lining |
| Material | Leather |
| Size | 39 |
| Sole Features | Removable, textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole. |
| Sole Material | Polyurethane, Composite Fabric |
| Style | Low |
| Toe Material | Composite |
| Weight (Metric) | 563 g per shoe in size 8 |
| Certifications/Compliance (Europe) | EN ISO 20345 : 2011 |