Learn More
Eppendorf™ Freezer Rack: Innova™ U101, CryoCube™ F101h, 3 in/76 mm
Description
Freezer Rack: Innova™ U101, CryoCube™ F101h, 3 in/76 mm, drawer, stainless steel- Rack for 76 mm/3 in boxes- Max. box size [mm]: 136 x 136- Drawer rack- Ext. dimensions of rack (DxWxH) [mm]: 308 x 145 x 244 (12.1 x 5.7 x 9.6 in)- Max. box number per rack: 6 (3 x 2)- Stainless steel- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 6 racks)- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 36 boxes)
Specifications
Specifications
| Product Type | Freezer Rack |