Learn More
Eppendorf™ Freezer Rack: Innova™ U360, U535, U725/-G, CryoCube™ F740 series (3-comp., Std),
Product Code. 15773235
Product Code. 15773235
Brand: Eppendorf™ 6001012310
Only null left
Add to basket
Add to basket
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Innova™ U360, U535, U725, U725-G, CryoCube™ F740 series (3-compartment, Standard), 3 in/76 mm, drawer, stainless steel- Rack for 76 mm/3 in boxes- Max. box size [mm]: 136 x 136- Drawer rack- Ext. dimensions of rack (DxWxH) [mm]: 580 x 140 x 414 (22.8 x 5.5 x 16.3 in)- Max. box number per rack: 20 (4 x 5)- Stainless steel- For ULT freezer: Innova U360 (max. 9 racks), Innova U535 (max. 12 racks), Innova U725 and U725-G (max. 18 racks), CryoCube F740 series (3-compartment version; max. 18 racks)- For ULT freezer: Innova U360 (max. 180 boxes), Innova U535 (max. 240 boxes), Innova U725 and 725-G (max. 360 boxes), CryoCube F740 series (3-compartment version; max. 360 boxes)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack