missing translation for 'onlineSavingsMsg'
Learn More

Frentizole, MedChemExpress

Product Code. 16436478
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16436478 10 mg 10mg
16409067 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16436478 Supplier Medchem Express Supplier No. HY15374

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Frentizole, an FDA-approved immunosuppressive drug, is a novel inhibitor of the A(beta)-ABAD interaction.

Frentizole, an FDA-approved immunosuppressive drug, is a novel inhibitor of the A(beta)-ABAD interaction.

Specifications

Chemical Name or Material Frentizole
CAS 26130-02-9
Color White
Purity Grade Notes Research
Molecular Formula C15H13N3O2S
Quantity 10 mg
Solubility Information DMSO : 12.5 mg/mL (41.76 mM; Need ultrasonic)
SMILES O=C(NC1=CC=CC=C1)NC2=NC3=CC=C(OC)C=C3S2
Molecular Weight (g/mol) 299.35
Formula Weight 299.35
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.