missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frequenin Antibody (11B8), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59480AF594
This item is not returnable.
View return policy
Description
Frequenin Monoclonal antibody specifically detects Frequenin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| Frequenin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| DKFZp761L1223, FREQFLUP, frequenin (Drosophila) homolog, Frequenin homolog, frequenin homolog (Drosophila), Frequenin-like protein, Frequenin-like ubiquitous protein, NCS-1, neuronal calcium sensor 1 | |
| Recombinant human Frequenin (1-190aa) purified from E. coli | |
| 0.1 mL | |
| Neuroscience | |
| 23413 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA | |
| 11B8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction