Learn More
Invitrogen™ Fresolimumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559353
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Fresolimumab has been used in trials studying the treatment of Primary Brain Tumors, Metastatic Breast Cancer, Diffuse Systemic Sclerosis, Pleural Malignant Mesothelioma, and Primary Focal Segmental Glomerulosclerosis.
Specifications
| Fresolimumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GC-1008 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4SP |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |