Learn More
Invitrogen™ Frg1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA570039
Product Code. 15898443
  • € 520.00 / 100µL
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
This gene maps to a location 100 kb centromeric of the repeat units on chromosome 4q35 which are deleted in facioscapulohumeral muscular dystrophy . It is evolutionarily conserved and has related sequences on multiple human chromosomes but DNA sequence analysis did not reveal any homology to known genes. In vivo studies demonstrate the encoded protein is localized to the nucleolus.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Frg1
Polyclonal
Unconjugated
FRG1
facioscapulohumeral muscular dystrophy region gene-1; Frg1; FRG1A; Frg1l1; FSG1; FSHD region gene 1; FSHD region gene 1 protein; FSHD region gene 1-like 1; hypothetical protein LOC550491; protein FRG1; Unknown (protein for MGC:140313); zgc:112426
Rabbit
Affinity Chromatography
RUO
686103, 689484
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5-1.0 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
0
FRG1, Frg1l1
synthetic peptide corresponding to a region of rat Frg1.
100 μL
Primary
Rat