missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled-1 Antibody (162531) [Janelia Fluor™ 549], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB11201I
This item is not returnable.
View return policy
Description
Frizzled-1 Monoclonal antibody specifically detects Frizzled-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| Frizzled-1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 162531 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| DKFZp564G072, FLJ95923, frizzled family receptor 1, frizzled homolog 1, frizzled, Drosophila, homolog of, 1, frizzled-1, fz-1, fzE1, hFz1, seven transmembrane spanning receptor, Wnt receptor | |
| Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-1, Gln72-His248 (Met122Ile), Accession # O70421 | |
| 0.1 mL | |
| GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway | |
| 8321 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction