missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled-4 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-93397
This item is not returnable.
View return policy
Description
Frizzled-4 Polyclonal specifically detects Frizzled-4 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Frizzled-4 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CD344, CD344 antigen, EVR1, exudative vitreoretinopathy 1, FEVR, frizzled (Drosophila) homolog 4, frizzled homolog 4 (Drosophila), frizzled-4, Fz-4, FZD4S, FzE4, GPCR, hFz4, MGC34390, WNT receptor frizzled-4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_036325.2 | |
| FZD4 | |
| Peptide with sequence DEEERRCDPIRIS corresponding to internal region according to NP_036325.2. | |
| 0.1 mg | |
| Wnt Signaling Pathway | |
| 8322 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction