Learn More
Novus Biologicals™ Frizzled-7 Antibody (U.Toronto patent anti-FZD7) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28845-50ug
 View more versions of this product
Product Code. 30045729
  • € 213.00 / 50µg
Description
Frizzled-7 Monoclonal antibody specifically detects Frizzled-7 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Frizzled-7
Monoclonal
Unconjugated
O75084
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
U.Toronto patent anti-FZD7
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
frizzled (Drosophila) homolog 7, frizzled homolog 7 (Drosophila), Frizzled, drosophila, homolog of, 7, Fz-7, FzE3frizzled-7, hFz7
FZD7
50 μg
GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway
8324