missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frk Antibody (OTI6D2), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70775APC
This item is not returnable.
View return policy
Description
Frk Monoclonal specifically detects Frk in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Frk | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| FRK | |
| Full length recombinant protein of human Frk (NP_002022) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI6D2 | |
| Flow Cytometry | |
| EC 2.7.10, fyn-related kinaseProtein-tyrosine kinase 5, GTKEC 2.7.10.2, Nuclear tyrosine protein kinase RAK, PTK5 protein tyrosine kinase 5, RAKPTK5, tyrosine-protein kinase FRK | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 2444.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction