missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRK Monoclonal Antibody (OTI3E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501052
This item is not returnable.
View return policy
Description
FRK (Fyn-related kinase), also known as Rak. It is an epithelial tissue-specific kinase. The human FRK gene maps to chromosome 6q21-q22.3 and encodes a 505 amino acid protein. The protein belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth.Specifications
| FRK | |
| Monoclonal | |
| 0.92 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P42685 | |
| FRK | |
| Full length human recombit protein of human FRK produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI3E2 | |
| Unconjugated | |
| FRK | |
| GTK, PTK5, RAK | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2444 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction