missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRMD3, Mouse anti-Human, Clone: 2F1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00257019-M09.100ug
This item is not returnable.
View return policy
Description
Sequence: SAPLISSSPVKAAREYEDPPSEEEDKIKEEPLTISELVYNPSASLLPTPVDDDEIDMLFDCPSRLELEREDTDSFEDLEADENAFLIASpecifications
| FRMD3 | |
| Monoclonal | |
| Unconjugated | |
| FERM domain containing 3 | |
| 4.1O/EPB41L4O/EPB41LO/MGC20553/P410 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2F1 | |
| In 1x PBS, pH 7.4 | |
| NM_174938 | |
| FRMD3 | |
| FRMD3 (NP_777598.2, 393 a.a. ∼ 480 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 257019 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction