missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FRMD4B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552677
This item is not returnable.
View return policy
Description
Immunogen sequence: LRGWYQRASG QKDQGHSPQT SFDSDRGSQR CLGFAGLQVP CSPSSRASLY SSVSSTNASG NWRTQLTIGL SDYETPAHSS YTSCYGNVYN PLPSPSRQYT EISQLDGTDG N Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.
Specifications
| FRMD4B | |
| Polyclonal | |
| Unconjugated | |
| FRMD4B | |
| 6030440G05Rik; C87375; FERM domain containing 4B; FERM domain-containing protein 4B; Frmd4b; GOBLIN; Golgi-associated band 4.1-like protein; GRP1 binding protein GRSP1; GRP1-binding protein GRSP1; GRSP1; KIAA1013; mKIAA1013; Nitzin; R74720 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23150 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2L6 | |
| FRMD4B | |
| Recombinant protein corresponding to Human FRMD4B. Recombinant protein control fragment (Product #RP-90636). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction