missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRMD6 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18421R
This item is not returnable.
View return policy
Description
FRMD6 Polyclonal antibody specifically detects FRMD6 in Human, Mouse samples. It is validated for Western Blot
Specifications
| FRMD6 | |
| Polyclonal | |
| Western Blot | |
| FERM domain containing 6 | |
| Recognizes region between residues Between Between 572 and 622.. | |
| 0.1 mL | |
| Cancer, Cell Biology | |
| 122786 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction