Learn More
Wesbart™ Front Access Freezer Racks
Click to view available options
Dimensions (L x W x H):
560 x 140 x 415 mm
560 x 140 x 420 mm
560 x 145 x 235 mm
560 x 280 x 290 mm
590 x 165 x 420 mm
675 x 140 x 305 mm
Positions:
2
3
4
7
8
10
Unit Size:
Each
Description
- ULT freezer racks
- For upright freezers to store boxes
Specifications
Specifications
| Includes | 4 Trays |
| Product Type | Freezer Rack |
| Description | Aluminum Front Access Rack |
| Autoclavable | Autoclavable |
| Color | Silver |
| Positions | 4 |
| Shape | Rectangular |
| Stackable | Yes |
| Sterility | Non-sterile |
| For Use With (Application) | Store Loose Items |
| Show More |