Learn More
Amarell™ Standard Milk Glass Total Immersion Thermometer
Product Code. 10110531
Change view
Click to view available options
Temperature Range (Metric):
-10°C to +100°C
-10°C to +150°C
-10°C to +200°C
-10°C to +250°C
-35°C to +50°C
Thermometer Fill:
Blue Liquid
Red Liquid
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Thermometer Fill Unit Size
10110531 -10°C to +100°C Red Liquid Each
10350231 -35°C to +50°C Red Liquid Each
10652621 -10°C to +200°C Red Liquid Each
10323911 -10°C to +250°C Blue Liquid Each
10232661 -10°C to +150°C Red Liquid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10110531 Supplier Amarell™ Supplier No. G10006FL
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use for total immersion applications. The standard milk glass total immersion thermometers use non-mercury colored fill.
  • Colorless, capillary prismatic
  • Total immersion
  • Diameter: 7.0 to 8.0mm
  • In square plastic case
Specifications
Immersion Type Total Immersion
Length (Metric) 260 mm
Material Glass
Thermometer Fill Red Liquid
Product Type Total Immersion Thermometer
Max. Temperature (Metric) 100°C
Min. Temperature (Metric) -10°C
Temperature Range (Metric) -10°C to +100°C
Subdivision 1°C