Learn More
Amarell™ Standard Milk Glass Total Immersion Thermometer
Use for total immersion applications. The standard milk glass total immersion thermometers use non-mercury colored fill.
Brand: Amarell™ G10310
Additional Details : Weight : 0.02200kg
Description
- Colorless, capillary prismatic
- Total immersion
- Diameter: 7.0 to 8.0mm
- In square plastic case
Specifications
Total Immersion | |
Glass | |
Total Immersion Thermometer | |
-10°C | |
1°C |
260 mm | |
Red Liquid | |
150°C | |
-10°C to +150°C |