missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPEX™ Frosted PE Vial Set
Shop All SPEX Certiprep Products
Click to view available options
Packaging Quantity:
24 Vials
Unit Size:
10 sets
Description
- Makes sample preparation faster and easier
- Unique alternative to standard titer plates
- Each grinding vial has a volume of 5 mL and a working volume of 1.5 mL
- Grinding vials are extensively tested with various samples and shown to resist perforation by steel grinding balls
- Polyethylene is compatible with commonly used reagents such as TRIzol™
- Frosting on the outside of the vial facilitates vial labeling
- Grinding vials are compatible with 2663 Cryo-Block to preserve temperature sensitive samples during grinding
- Process 96 samples at a time by running four Vial Sets in the Large Capacity Clamp Assembly
Specifications
Specifications
| Capacity (Metric) | 5 mL |
| Diameter (Metric) | 9.5 mm |
| Material | Frosted Polyethylene |
| Packaging Quantity | 24 Vials |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction