missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRS2 Antibody (462902), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB40691MFV450
This item is not returnable.
View return policy
Description
FRS2 Monoclonal antibody specifically detects FRS2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| FRS2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 462902 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| FGFR signalling adaptor, FGFR-signaling adaptor SNT, fibroblast growth factor receptor substrate 2, FRS2A, FRS2alpha, SNT, SNT1, SNT-1FGFR substrate 2, Suc1-associated neurotrophic factor target 1 | |
| E. coli-derived recombinant human FRS2, Asn121-Asn449, Accession # Q8WU20 | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 10818 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction