missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRS2 Antibody (462902), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB40691C
This item is not returnable.
View return policy
Description
FRS2 Monoclonal antibody specifically detects FRS2 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| FRS2 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 462902 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| FGFR signalling adaptor, FGFR-signaling adaptor SNT, fibroblast growth factor receptor substrate 2, FRS2A, FRS2alpha, SNT, SNT1, SNT-1FGFR substrate 2, Suc1-associated neurotrophic factor target 1 | |
| E. coli-derived recombinant human FRS2, Asn121-Asn449, Accession # Q8WU20 | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 10818 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction