missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fructosyl amino acid oxidase Antibody, Alexa Fluor™ 594, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP1-77782AF594
This item is not returnable.
View return policy
Description
Fructosyl amino acid oxidase Polyclonal antibody specifically detects Fructosyl amino acid oxidase in Bacteria samples. It is validated for Western Blot, ELISA
Specifications
| Fructosyl amino acid oxidase | |
| Polyclonal | |
| Western Blot, ELISA | |
| EC=1.5.3.-, FAOX-C | |
| Fructosyl amino acid oxidase [from Corynebacterium sp. Expressed in E.coli] (Uniprot: Q8RIU8) | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. |
| Western Blot, ELISA | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Sheep | |
| Multi-step | |
| RUO | |
| Bacteria | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction