missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FRYL Polyclonal Antibody
GREENER_CHOICE

Product Code. 15764551 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15764551 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15764551 Supplier Invitrogen™ Supplier No. PA556644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: AYCKLINQVN TIKNEAEVIN MSEELAQLES ILKEAESASE NEEIDISKAA QTTIETAIHS LIETLKNKEF ISAVAQVKA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 92%.

FRYL gene ontology annotations related to this gene include cell morphogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRYL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FRYL
Gene Accession No. O94915
Gene Alias 2010313D22Rik; 2310004H21Rik; 2510002A14Rik; 9030227G01Rik; AF4p12; ALL1-fused gene from chromosome 4p12 protein; DKFZp686E205; FRY like; FRY like transcription coactivator; FRYL; FRY-like; furry homolog-like; furry homolog-like (Drosophila); furry-like; KIAA0826; mKIAA0826; MOR2; mor2 cell polarity protein homolog; protein furry homolog-like; RGD1311309
Gene Symbols FRYL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FRYL. Recombinant protein control fragment (Product #RP-95480).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285527, 364144, 72313
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.