missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSBA Antibody (FSBA 2F5/5), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2656C
This item is not returnable.
View return policy
Description
FSBA Monoclonal antibody specifically detects FSBA in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| FSBA | |
| Monoclonal | |
| DyLight 650 | |
| 5'-(4-(fluorosulfonyl)benzoyl)adenosine, 5'-(4-fluorosulfonylbenzoyl)adenosine, 5'-FSBA, p-fluorosulfonylbenzoyl-5'-adenosine | |
| Fluorylsulponylbenzoyladenosine (FSBA) | |
| 0.1 mL | |
| Epitope Tags | |
| Non-species specific | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| FSBA 2F5/5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction