Learn More
FSCN1 Monoclonal Antibody (OTI2C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807294
Encompass_Preferred
 View more versions of this product
Product Code. 17587703
  • € 540.87 / 100µL
Description
Human fascin is a highly conserved actin-bundling protein. Fascin, encoded by the human homolog for sn (hsn) gene, has been localized to microspikes and stress fibers of cultured cells where it is thought to be involved in the formation of microfilament bundles. It is expressed predomitly in dendritic cells. Lymphoid cells, myeloid cells and plasma cells are negative. However. Reed Sternberg cells in Hodgkin′s lymphoma are positive for fascin staining. Epstein-Barr virus may induce expression of fascin in B cells.
Specifications
Specifications
FSCN1
Monoclonal
Unconjugated
FSCN1
FAN1, HSN, SNL, p55
Mouse
Affinity Chromatography
RUO
6624
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI2C3
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q16658
FSCN1
Full length human recombit protein of human FSCN1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b