missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSD1, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00079187-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MEEQREALRKIIKTLAVKNEEIQSFIYSLKQMLLNVEANSAKVQEDLEAEFQSLFSLLEELKEGMLMKIKQDRASRTYELQNQLAACTRALESSEELLETANQTLQAMDSEDFPQAAKQIKDGVTMAPAFRLSLKAKVSDNMSHLMVDFAQERQMLQALKFLPVPSAPVIDLAESLVADNCVTLVWRMPDEDSKIDHYVLEYRRTNFEGPPRLKEDQPWMVIEGIRQTEYTLTGLKFDMKYMNFRVKACNKAVAGEFSEPVTLETPAFMFRLDASTSHQNLRVDDLSVEWDAMGGKVQDIKAREKDGKGRTASPINSPARGTPSPKRMPSGRGGRDRFTAESYTVLGDTLIDGGEHYWEVRYEPDSKAFGVGVAYRSLGRFEQLGKTAASWCLHVNNWLQVSFTAKHANKVKVLDAPVPDCLGVHCDFHQGLLSFYNARTKQVLHTFKTRFTQPLLPAFTVWCGSFQVTTGLQVPSAVRCLQKRGSATSSSNTSLTSpecifications
| FSD1 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_024333.1 | |
| FSD1 | |
| FSD1 (NP_077309.1, 1 a.a. ∼ 496 a.a) full-length human protein. | |
| RUO | |
| 79187 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| fibronectin type III and SPRY domain containing 1 | |
| GLFND/MGC3213/MIR1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido