missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSH beta Antibody (OTI2A6), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70781AF488
This item is not returnable.
View return policy
Description
FSH beta Monoclonal specifically detects FSH beta in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| FSH beta | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FSHB | |
| Full length recombinant protein of human FSH beta (NP_001018090) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI2A6 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| follicle stimulating hormone, beta polypeptide, Follicle-stimulating hormone beta subunit, Follitropin beta chain, follitropin subunit beta, follitropin, beta chain, FSH-B, FSH-beta | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 2488.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction