missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FSH beta Polyclonal Antibody
GREENER_CHOICE

Product Code. 17199563
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17199563 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17199563 Supplier Invitrogen™ Supplier No. PA5111651

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TRDLVYKDPA RPKIQKTCTF KELVYETVRV PGCAHHADSL YTYPVATQCH CGK Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 85%.

Human follicle stimulating hormone is a glycoprotein produced from urine of post-menopausal women and having a total molecular mass of 35,500 Dalton. HuFSH is a heterodimeric hormone consisting of 165 amino acids a chain (non-active) and 115 amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSH beta
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FSHB
Gene Accession No. P01225
Gene Alias follicle stimulating hormone beta subunit; follicle stimulating hormone, beta polypeptide; follicle-stimulating hormone beta subunit; Follitropin beta chain; follitropin subunit beta; follitropin, beta chain; FSHB; FSH-B; FSH-beta; HH24
Gene Symbols FSHB
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human FSH-beta. Recombinant protein control fragment (Product #RP-107705).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2488
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.