missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSHB Monoclonal Antibody (OTI4E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501623S
This item is not returnable.
View return policy
Description
FSHB Monoclonal antibody specifically detects FSHB in Canine, Human, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| FSHB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99MB4, Q9BRT8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 171057, 55871 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI4E10 | |
| Unconjugated | |
| CBWD1 | |
| class 1 follicle-stimulating hormone beta polypeptide; class 2 follicle-stimulating hormone beta polypeptide; follicle - stimulating hormone subunit beta; follicle stimulating hormone beta; follicle stimulating hormone beta subunit; follicle stimulating hormone beta-subunit precursor; follicle stimulating hormone subunit beta; follicle stimulating hormone, beta polypeptide; follicle-stimulating hormone beta subunit; follicle-stimulating hormone beta-subunit; follicle-stimulating hormone subunit beta; follicule stimulating hormone, beta polypeptide; follitropin beta chain; Follitropin subunit beta; follitropin, beta chain; FSH; FSH-alpha; FSHB; FSH-B; Fshbeta; FSH-beta; HH24; LSH-alpha; prefollicle stimulating hormone beta chain; TSH-alpha | |
| Full length human recombinant protein of human CBWD1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction