missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSHR Antibody (626717), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB65591H
This item is not returnable.
View return policy
Description
FSHR Monoclonal antibody specifically detects FSHR in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF
Specifications
| FSHR | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Neutralization, CyTOF | |
| 626717 | |
| Flow Cytometry, Neutralization, CyTOF-ready | |
| follicle stimulating hormone receptor, follicle-stimulating hormone receptor, Follitropin receptor, FSH receptor, FSH-R, LGR1FSHRO, MGC141667, MGC141668, ODG1 | |
| NS0 mouse myeloma cell line transfected with human FSH R, Accession # M65085 | |
| 0.5 mL | |
| GPCR | |
| 2492 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction