missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSHR Polyclonal Antibody

FSHR Polyclonal Antibody
Brand: Invitrogen PA533548
This item is not returnable.
View return policy
Description
FSHR Polyclonal Antibody for IHC (P)
Specifications
| FSHR | |
| Polyclonal | |
| Unconjugated | |
| FSHR | |
| follicle stimulating hormone receptor; follicle-stimulating hormone receptor; Follitropin receptor; FSH receptor; Fshr; FSH-R; FSHR1; FSHRO; LGR1; ODG1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100052957, 100343287, 100861291, 2492, 25449, 281172, 397679, 443299 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P20395, P23945, P35376, P35379, P47799, P49059 | |
| FSHR | |
| Synthetic 18 amino acid peptide from N-terminal extracellular domain of human FSHR. | |
| 50 μg | |
| Primary | |
| Human, Rat, Rabbit, Monkey, Bovine, Sheep, Pig, Horse, Goat, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction