missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSIP1 Antibody (OTI4F9), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72451JF549
This item is not returnable.
View return policy
Description
FSIP1 Monoclonal specifically detects FSIP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| FSIP1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FSIP1 | |
| Full length recombinant protein of human FSIP1 (NP_689810) produced in HEK293T cells. | |
| 0.1 mL | |
| 161835.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4F9 | |
| Western Blot, Immunohistochemistry | |
| fibrous sheath interacting protein 1, fibrous sheath-interacting protein 1, FLJ35989 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction