missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCSK9 Monoclonal Antibody (OTI8C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811842
This item is not returnable.
View return policy
Description
This gene encodes a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. The encoded protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. This protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons. Mutations in this gene have been associated with a third form of autosomal domit familial hypercholesterolemia.Specifications
| PCSK9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NBP7 | |
| PCSK9 | |
| Human recombit protein fragment corresponding to amino acids 153-692 of human PCSK9 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI8C12 | |
| Unconjugated | |
| PCSK9 | |
| FH3, HCHOLA3, LDLCQ1, NARC-1, NARC1, PC9 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 255738 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction