missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: WMDPGTSRGP DVGVGESQAE EPRSFEVTRR EGLSSHNELL ASCGKKFCSR GSRCVLSRKT GEPECQCLEA CRPSYVPVCG S Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 65%.
FSTL4 (Follistatin Like 4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includecalcium ion binding. An important paralog of this gene isFSTL5.
Specifications
Specifications
| Antigen | FSTL4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FSTL4 |
| Gene Accession No. | Q6MZW2 |
| Gene Alias | B230374F23Rik; follistatin like 4; follistatin-like 4; follistatin-like protein 4; Follistatin-related protein 4; FSL4; FSTL4; KIAA1061; m-D/Bsp120I 1-1; mKIAA1061; SPIG1 |
| Gene Symbols | FSTL4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
