missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FT4 Competitive ELISA Kit

Product Code. 18027454
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 18027454

Brand: Invitrogen™ EEL133

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Free Thyroxine (FT4) is a hormone produced by the thyroid gland and plays a crucial role in regulating metabolism, growth, and development in the body. It is transported in the bloodstream and acts on target cells by binding to thyroid hormone receptors. The structure of FT4 is characterized by a tyrosine residue linked to an iodine atom, forming the thyroid hormone. It has a molecular weight of 777.00 g/mol and a chemical formula of C15H11I4NO4. FT4 is primarily produced in the thyroid gland and is regulated by the hypothalamic-pituitary-thyroid axis. Its levels are typically measured in blood tests to diagnose thyroid disorders.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.56 to 100 pg/mL
Assay Sensitivity 0.94 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target FT4
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.