Learn More
FTCD Monoclonal Antibody (OTI4A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505028S
Encompass_Preferred
 View more versions of this product
Product Code. 17522962
  • € 242.00 / 30µL
Description
FTCD Monoclonal antibody specifically detects FTCD in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
FTCD
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P62508
Mouse
Affinity chromatography
RUO
2104
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
OTI4A7
Unconjugated
Esrrg
58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1
Human recombinant protein fragment corresponding to amino acids 24-263 of human ESRRG produced in E.coli.
30 μL