missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTCD Monoclonal Antibody (OTI4F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504951
This item is not returnable.
View return policy
Description
Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.Specifications
| FTCD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95954 | |
| FTCD | |
| Full length human recombit protein of human FTCD produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F5 | |
| Unconjugated | |
| FTCD | |
| LCHC1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10841 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction