Learn More
FTO Antibody (5-2H10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-29512
Product Code. 18728233
  • € 482.00 / 0.10mL
Description
FTO Monoclonal specifically detects FTO in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FTO
Monoclonal
Unconjugated
Q9C0B1
FTO
Synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH. Accession # Q9C0B1
Affinity Purified
RUO
Primary
Specific for the ∼58 kDa FTO protein in Western blots of rat testes lysate. The antibody has also been used for immunocytochemistry with neuronal progenitor cells.
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, Immunocytochemistry, Immunofluorescence
5-2H10
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100
alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149
Mouse
58 kDa
0.1 mL
Lipid and Metabolism, Neuroscience
79068