missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTO Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77021AF405
This item is not returnable.
View return policy
Description
FTO Polyclonal antibody specifically detects FTO in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| FTO | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human FTO. The immunogen is located within the first 50 amino acids of FTO. Amino Acid Squence: TWLPYLTPKDDEFYQ | |
| 0.1 mL | |
| Lipid and Metabolism, Neuroscience | |
| 79068 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction