missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTO Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-60935B
This item is not returnable.
View return policy
Description
FTO Polyclonal specifically detects FTO in Human, Mouse, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoblotting.
Specifications
| FTO | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunoblotting | |
| Q9C0B1 | |
| FTO | |
| A synthetic peptide within the C-terminal region (residues 400-505) of the human FTO protein. [Swiss-Prot# Q9C0B1] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Chicken, Primate | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoblot | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
| Rabbit | |
| 60 kDa | |
| 0.1 mL | |
| Lipid and Metabolism, Neuroscience | |
| 79068 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction