missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTO Antibody (FT86-4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-80045CL1
This item is not returnable.
View return policy
Description
FTO Monoclonal antibody specifically detects FTO in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| FTO | |
| Monoclonal | |
| CoraFluor 1 | |
| alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
| Recombinant human FTO. | |
| 0.1 mL | |
| Lipid and Metabolism, Neuroscience | |
| 79068 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| FT86-4 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction