missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FTO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584785
This item is not returnable.
View return policy
Description
Immunogen sequence: LFRDLVRIQG KDLLTPVSRI LIGNPGCTYK YLNTRLFTVP WPVKGSNIKH TEAEIAAACE TFLKLNDYLQ IETIQALEEL AA Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 88%.
FTO is a novel protein and a member of the non-heme dioxygenase (Fe(II)-and 2-oxoglutarate-dependent dioxygenases) superfamily. Amino acid conservation patterns support this hypothesis and indicates that both 2-oxoglutarate and iron should be important for FTO function. Though not much is yet known about FTO, it is thought that the protein may somehow modify the activity of genes involved in metabolism and fat storage, which in turn may influence a person's risk of obesity.
Specifications
| FTO | |
| Polyclonal | |
| Unconjugated | |
| Fto | |
| AlkB homolog 9; ALKBH9; alpha-ketoglutarate-dependent dioxygenase FTO; AW743446; BMIQ14; fat mass and obesity associated; fat mass and obesity-associated protein; fatso; FTO; GDFD; KIAA1752; m6A(m)-demethylase FTO; MGC5149; mKIAA1752; mRNA (2'-O-methyladenosine-N(6)-)-demethylase FTO; mRNA N(6)-methyladenosine demethylase FTO; Protein fatso; protein fto; RGD1305121; tRNA N1-methyl adenine demethylase FTO; U6 small nuclear RNA (2'-O-methyladenosine-N(6)-)-demethylase FTO; U6 small nuclear RNA N(6)-methyladenosine-demethylase FTO | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79068 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9C0B1 | |
| Fto | |
| Recombinant protein corresponding to Human FTO. Recombinant protein control fragment (Product #RP-108242). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction